Learn More
Alkali Scientific CellPro 15mL Bio-Reaction Tube
SDP
Catalog No. 503026512
Encompass_Preferred
Our 15mL Bio-Reaction Tubes provide an economical and efficient solution for small-scale high-density suspension cell culture and other biological reaction applications Designed with a 0 22 m hydrophobic vented membrane these tubes ensure free gas exchange while maintaining aseptic conditions making them ideal for a variety of laboratory workflows Optimized for Suspension Cell Culture Supports high-density cell growth in a controlled environment Reliable Aseptic Conditions Ultrasonically welded 0 22 m hydrophobic membrane enables sterile gas exchange Versatile Applications Ideal for suspension cell culture bacterial growth seeding larger bioreactors and lyophilization Efficient Screening and Optimization Allows researchers to optimize reaction parameters using small amounts of material enabling multiple parallel experiments Dual-Purpose Design Enables both culturing and centrifugation in a single tube minimizing contamination risks and streamlining workflows High-Quality Po
Catalog No. 50-302-6512 Supplier Alkali Scientific Supplier No. CB5601
Our 15mL Bio-Reaction Tubes provide an economical and efficient solution for small-scale high-density suspension cell culture and other biological reaction applications Designed with a 0 22 m hydrophobic vented membrane these tubes ensure free gas exchange while maintaining aseptic conditions making them ideal for a variety of laboratory workflows Optimized for Suspension Cell Culture Supports high-density cell growth in a controlled environment Reliable Aseptic Conditions Ultrasonically welded 0 22 m hydrophobic membrane enables sterile gas exchange Versatile Applications Ideal for suspension cell culture bacterial growth seeding larger bioreactors and lyophilization Efficient Screening and Optimization Allows researchers to optimize reaction parameters using small amounts of material enabling multiple parallel experiments Dual-Purpose Design Enables both culturing and centrifugation in a single tube minimizing contamination risks and streamlining workflows High-Quality Po