Learn More
Alkali Scientific CellPro™ 1X DPBS, Dulbecco’s Phosphate Buffered Saline, without Calcium or Magnesium, Sterile Filtered, 500mL, 10/Case
SDP
Catalog No. 503029090
Encompass_Preferred
Like PBS, DPBS is also a water-based solution containing sodium chloride and phosphate buffer that is used in biological applications. However, DPBS differs from PBS in that it also includes potassium chloride and is available in a larger variety of formulations.
Catalog No. 50-302-9090 Supplier Alkali Scientific Supplier No. PBD500CS
Like PBS, DPBS is also a water-based solution containing sodium chloride and phosphate buffer that is used in biological applications. However, DPBS differs from PBS in that it also includes potassium chloride and is available in a larger variety of formulations.