Learn More
Alkali Scientific CellPro™ 1X DPBS, Dulbecco’s Phosphate Buffered Saline, without Calcium or Magnesium, Sterile Filtered, 1L, 10/Case
SDP
Catalog No. 503027632
Encompass_Preferred
Like PBS, DPBS is also a water-based solution containing sodium chloride and phosphate buffer that is used in biological applications. However, DPBS differs from PBS in that it also includes potassium chloride and is available in a larger variety of formulations.
Catalog No. 50-302-7632 Supplier Alkali Scientific Supplier No. PBD1000CS
Add to Cart
Add to Cart
Like PBS, DPBS is also a water-based solution containing sodium chloride and phosphate buffer that is used in biological applications. However, DPBS differs from PBS in that it also includes potassium chloride and is available in a larger variety of formulations.