Learn More
Alkali Scientific CellPro™ 2.0 ml 96-Well Deep Well Plate, U-Bottom, Round Well, 5/pk, 50/cs, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503029441
Encompass_Preferred
Heavy polypropylene 96-Well Deep Well Plate printed alphanumeric labeling, and smooth interior surfaces to ensure that all the sample can be pipetted. Minimal binding / liquid retention Black printed alpha-numerics* Virgin, medical-grade polypropylene 3-Sided labeling and SBS footprint for ease of use on robotic systems Manufactured with 30% solar power and medical grade plastics Autoclavable Packing: 10pk, 50/cs.  
Catalog No. 50-302-9441 Supplier Alkali Scientific Supplier No. MPD35
Heavy polypropylene 96-Well Deep Well Plate printed alphanumeric labeling, and smooth interior surfaces to ensure that all the sample can be pipetted. Minimal binding / liquid retention Black printed alpha-numerics* Virgin, medical-grade polypropylene 3-Sided labeling and SBS footprint for ease of use on robotic systems Manufactured with 30% solar power and medical grade plastics Autoclavable Packing: 10pk, 50/cs.  