Learn More
Alkali Scientific CellPro™ 2.0mL Microcentrifuge Tubes, Lock Cap, Clear, Round Bottom, DNase, RNase & Pyrogen Free, 500/Pk, 5000/Cs
SDP
Catalog No. 503029400
Encompass_Preferred
Our centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. USP VI grade polypropylene material, free from heavy metal. RCF: 30,000 xg. Special safe-lock cap design provides secure lid sealing. Clear graduations and frosted marking area. Easy to open and close. Autoclavable. RNase free, endotoxin level less than 0.1EU.
Catalog No. 50-302-9400 Supplier Alkali Scientific Supplier No. CN3020
Our centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. USP VI grade polypropylene material, free from heavy metal. RCF: 30,000 xg. Special safe-lock cap design provides secure lid sealing. Clear graduations and frosted marking area. Easy to open and close. Autoclavable. RNase free, endotoxin level less than 0.1EU.