Learn More
Alkali Scientific CellPro™ 2.0mL Screw Tubes, conical base, non-Graduated, ribbed, Clear, 500/pk, 5000/case
SDP
Catalog No. 503026623
Encompass_Preferred
Volume: 2.0 mLGraduations: Non-GraduatedRibbing (Knurls): RibbedBase: ConicalColor: -00 ClearPackaging: 500/unit, 10 units/case
Catalog No. 50-302-6623 Supplier Alkali Scientific Supplier No. C3025
Volume: 2.0 mLGraduations: Non-GraduatedRibbing (Knurls): RibbedBase: ConicalColor: -00 ClearPackaging: 500/unit, 10 units/case