Learn More
Alkali Scientific CellPro™ 2.0mL, Sterile Microcentrifuge Tubes w/ O-ring & attached colorless screw caps, generic version without graduations, ideal for OEM packaging and reagent kits, 50/Bag, 1000/Case
SDP
Catalog No. 503029465
Encompass_Preferred
Leak-proof O-ring seal Ultra-clear polypropylene Packaged in resealablebags of 50 Sterile; RNase, DNase & Pyrogen free  Available with white, printed graduations CellPro™ screw-cap microtubes come sterile, in a variety of sizes, cap configurations, graduation options, and even a loop-cap option for the 2.0mL version. Made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -86°C to +121°C. All CellPro™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.
Catalog No. 50-302-9465 Supplier Alkali Scientific Supplier No. MT3170
Leak-proof O-ring seal Ultra-clear polypropylene Packaged in resealablebags of 50 Sterile; RNase, DNase & Pyrogen free  Available with white, printed graduations CellPro™ screw-cap microtubes come sterile, in a variety of sizes, cap configurations, graduation options, and even a loop-cap option for the 2.0mL version. Made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -86°C to +121°C. All CellPro™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.