Learn More
Alkali Scientific CellPro™ 24-Well, Silicone Sealing Mats, 50/pk, Sterile
SDP
Catalog No. 503027765
Encompass_Preferred
CellPro™ 24 Well, Silicone Sealing Mats seals 24 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.
Catalog No. 50-302-7765 Supplier Alkali Scientific Supplier No. SSM332021S
CellPro™ 24 Well, Silicone Sealing Mats seals 24 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.