Learn More
Alkali Scientific CellPro™ 24-Well, Silicone Sealing Mats, 50/pk, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503028325
Encompass_Preferred
CellPro™ 24 Well, Silicone Sealing Mats seals 24 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.
Catalog No. 50-302-8325 Supplier Alkali Scientific Supplier No. SSSM332021
CellPro™ 24 Well, Silicone Sealing Mats seals 24 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.