Learn More
Alkali Scientific CellPro™ 25cm2 Cell Culture Flask, Vent Cap, TC, Sterile 10/pk, 200/cs
SDP
Catalog No. 502336241
Encompass_Preferred
CellPro™ Vented TC- Treated Cell Culture Flasks are packaged in zip bags. They are high clarity, 100% virgin polystyrene, stackable, sterilized by E-Beam, and have frosted writing and clear graduations. They have vent caps with breathable 0.22um membrane, clear lot number for batch traceability and are non-pyrogenic, DNase/RNase free.
Catalog No. 50-233-6241 Supplier Alkali Scientific Supplier No. TVN0025
CellPro™ Vented TC- Treated Cell Culture Flasks are packaged in zip bags. They are high clarity, 100% virgin polystyrene, stackable, sterilized by E-Beam, and have frosted writing and clear graduations. They have vent caps with breathable 0.22um membrane, clear lot number for batch traceability and are non-pyrogenic, DNase/RNase free.