Learn More
Alkali Scientific CellPro 25mL Serological Pipets, Individually Wrapped, Sterile in resealable bulk bags 200/cs
SDP
Supplier:  Alkali Scientific SP225B
Encompass_Preferred
CellPro™ Serological Pipets are sterile and individually wrapped. They have color coded paper/plastic wrapping for pop-through or peel-apart techniques. Designed with quality in mind with a stretched, non-welded design and bi-directional and extra negative graduation marks. They contain a filter plug, are 100% polystyrene, and are Gamma sterilized as well as pyrogen, cytotoxin, RNase and Dnase free.
Catalog No. 50-233-6201