Learn More
Alkali Scientific CellPro™ 2X YT Broth 1L PC
SDP
Catalog No. 502389026
Encompass_Preferred
Alkali Scientific™ CellPro™ 2X YT Broth is a specialized growth medium optimized for the high-density culture of Escherichia coli strains. With its enriched formulation containing tryptone, yeast extract, and glycerol, this broth supports enhanced bacterial growth and recombinant protein expression. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for molecular biology and biotechnology applications.
Catalog No. 50-238-9026 Supplier Alkali Scientific Supplier No. DC0040
Alkali Scientific™ CellPro™ 2X YT Broth is a specialized growth medium optimized for the high-density culture of Escherichia coli strains. With its enriched formulation containing tryptone, yeast extract, and glycerol, this broth supports enhanced bacterial growth and recombinant protein expression. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for molecular biology and biotechnology applications.