Learn More
Alkali Scientific CellPro 3-Layer Cell Culture M
SDP
Catalog No. 503027144
Encompass_Preferred
The CellPro3-Layer Multi-Layer Cell Culture Flask provides 525 cm of total growth surface area offering an efficient and scalable solution for adherent cell culture Designed for uniform cell growth and optimal productivity these flasks enable consistent medium distribution across all layers enhancing cell expansion while minimizing handling time Manufactured from high-quality polystyrene (GPPS) with vacuum plasma tissue culture (TC) treatment the flask ensures superior cell adhesion Its leak-proof design prevents cross-contamination allowing for safe and efficient cell culture workflows Compatible with 10 mL serological pipettes the flask supports easy liquid aspiration replenishment and cell harvesting directly within the system Each flask is individually lot-numbered for traceability and conforms to USP Class VI standards Equipped with a vent cap made from high-density polyethylene (HDPE) and a polytetrafluoroethylene (PTFE) filter membrane the CellPro3-Layer Flask is steriliz
Catalog No. 50-302-7144 Supplier Alkali Scientific Supplier No. TV33L
The CellPro3-Layer Multi-Layer Cell Culture Flask provides 525 cm of total growth surface area offering an efficient and scalable solution for adherent cell culture Designed for uniform cell growth and optimal productivity these flasks enable consistent medium distribution across all layers enhancing cell expansion while minimizing handling time Manufactured from high-quality polystyrene (GPPS) with vacuum plasma tissue culture (TC) treatment the flask ensures superior cell adhesion Its leak-proof design prevents cross-contamination allowing for safe and efficient cell culture workflows Compatible with 10 mL serological pipettes the flask supports easy liquid aspiration replenishment and cell harvesting directly within the system Each flask is individually lot-numbered for traceability and conforms to USP Class VI standards Equipped with a vent cap made from high-density polyethylene (HDPE) and a polytetrafluoroethylene (PTFE) filter membrane the CellPro3-Layer Flask is steriliz