Learn More
Alkali Scientific CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/ Sterile/with Clear Cover
SDP
Catalog No. 5000662732
Encompass_Preferred
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile with Clear Cover. The high bind surface of this clear square ELISA plate ensures excellent protein or analyte binding for various assays. The clear cover provides protection, while the sterile packaging guarantees contamination-free use. Perfect for laboratory research, diagnostics, and clinical tests, the case contains 40 individually sealed plates. The product delivers accurate and reliable results, enhancing research outcomes.
Catalog No. 50-006-62732 Supplier Alkali Scientific Supplier No. DWP384CH
Add to Cart
Add to Cart
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile with Clear Cover. The high bind surface of this clear square ELISA plate ensures excellent protein or analyte binding for various assays. The clear cover provides protection, while the sterile packaging guarantees contamination-free use. Perfect for laboratory research, diagnostics, and clinical tests, the case contains 40 individually sealed plates. The product delivers accurate and reliable results, enhancing research outcomes.