Learn More
Alkali Scientific CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/ Sterile
SDP
Catalog No. 5000663310
Encompass_Preferred
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile. The high bind surface of this clear square ELISA plate makes it ideal for experiments requiring enhanced analyte binding. The plates are sterile and sealed to maintain their integrity during use. Designed to deliver reliable and reproducible results, this product is perfect for diagnostic and research applications. Each case includes 40 plates, and each plate is individually packaged to maintain sterility.
Catalog No. 50-006-63310 Supplier Alkali Scientific Supplier No. DWP384CHS
Add to Cart
Add to Cart
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile. The high bind surface of this clear square ELISA plate makes it ideal for experiments requiring enhanced analyte binding. The plates are sterile and sealed to maintain their integrity during use. Designed to deliver reliable and reproducible results, this product is perfect for diagnostic and research applications. Each case includes 40 plates, and each plate is individually packaged to maintain sterility.