Learn More
Alkali Scientific CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/DNase, RNase & Pyrogen Free/ with Clear Cover
SDP
Catalog No. 5000662733
Encompass_Preferred
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/DNase, RNase & Pyrogen Free with Clear Cover. This clear square ELISA plate is designed for critical assays where purity and high performance are needed. Free from DNase, RNase, and pyrogens, this plate ensures the integrity of sensitive experiments. It comes with a clear cover for protection and visibility. Each pack contains 10 plates, and the case holds 40 plates, all individually sealed for sterility.
Catalog No. 50-006-62733 Supplier Alkali Scientific Supplier No. DWP384CI
Add to Cart
Add to Cart
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/DNase, RNase & Pyrogen Free with Clear Cover. This clear square ELISA plate is designed for critical assays where purity and high performance are needed. Free from DNase, RNase, and pyrogens, this plate ensures the integrity of sensitive experiments. It comes with a clear cover for protection and visibility. Each pack contains 10 plates, and the case holds 40 plates, all individually sealed for sterility.