Learn More
Alkali Scientific CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/ Sterile/ with Clear Cover
SDP
Catalog No. 5000662004
Encompass_Preferred
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/Sterile with Clear Cover. This clear square plate features an intermediate binding surface, providing reliable performance in various ELISA assays. Sterile and individually sealed, the plates prevent contamination, ensuring accurate results. The clear design allows for easy sample monitoring and reliable data interpretation. Each case contains 40 plates, each packaged in a way to maintain sterility.
Catalog No. 50-006-62004 Supplier Alkali Scientific Supplier No. DWP384CIS
Add to Cart
Add to Cart
CellPro™ 384 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/Sterile with Clear Cover. This clear square plate features an intermediate binding surface, providing reliable performance in various ELISA assays. Sterile and individually sealed, the plates prevent contamination, ensuring accurate results. The clear design allows for easy sample monitoring and reliable data interpretation. Each case contains 40 plates, each packaged in a way to maintain sterility.