Learn More
Alkali Scientific CellPro™ 384-Well, Silicone Sealing Mats, 50/pk, 384 Square Well, Silicone, 50 / PK
SDP
Catalog No. 503028242
Encompass_Preferred
CellPro™ 384 Well, Silicone Sealing Mats seals 384 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.
Catalog No. 50-302-8242 Supplier Alkali Scientific Supplier No. SSM332041
CellPro™ 384 Well, Silicone Sealing Mats seals 384 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.