Learn More
Alkali Scientific CellPro™ 384 Well White Square ELISA Plate, 10/Pk, 40/Case, High Bind/ Sterile
SDP
Catalog No. 5000662341
Encompass_Preferred
CellPro™ 384 Well White Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile. This high bind 384-well ELISA plate is specifically designed for assays that require increased protein or analyte binding. Manufactured under strict sterility conditions, this plate ensures high-quality performance, offering consistent results for diagnostics and research. Each pack contains 10 plates, and every case includes 40 individually sealed plates.
Catalog No. 50-006-62341 Supplier Alkali Scientific Supplier No. DWP384WHS
Add to Cart
Add to Cart
CellPro™ 384 Well White Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile. This high bind 384-well ELISA plate is specifically designed for assays that require increased protein or analyte binding. Manufactured under strict sterility conditions, this plate ensures high-quality performance, offering consistent results for diagnostics and research. Each pack contains 10 plates, and every case includes 40 individually sealed plates.