Learn More
Alkali Scientific CellPro™ 384 Well White Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/ Sterile/ with Clear Cover
SDP
Catalog No. 5000663113
Encompass_Preferred
CellPro™ 384 Well White Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/Sterile, with Clear Cover. This 384-well plate is ideal for use in ELISA applications, providing reliable results in research and clinical diagnostics. The white square design with intermediate binding surface ensures optimal performance with various assay types. Manufactured by Alkali Scientific, each case contains 40 plates, each individually sealed to maintain sterility.
Catalog No. 50-006-63113 Supplier Alkali Scientific Supplier No. DWP384WIS
Add to Cart
Add to Cart
CellPro™ 384 Well White Square ELISA Plate, 10/Pk, 40/Case, Intermediate Bind/Sterile, with Clear Cover. This 384-well plate is ideal for use in ELISA applications, providing reliable results in research and clinical diagnostics. The white square design with intermediate binding surface ensures optimal performance with various assay types. Manufactured by Alkali Scientific, each case contains 40 plates, each individually sealed to maintain sterility.