Learn More
Alkali Scientific CellPro™ 48 Well Cell Culture Plate, Flat, Non-Treated, Sterile 1/pk, 50/cs
SDP
Catalog No. 502336197
Encompass_Preferred
Use Alkali Scientific Flat-Bottom Plates, Non-Treated for a variety of cell culture applications including suspension cell growth. These sterile plates have a clear, untreated, hydrophobic polystyrene surface, and are ready for use in an aseptic environment. These plates are RNase/DNase free and non-pyrogenic, and the included lid is non-reversible, designed to prevent cross-condensation between wells. Stackable for easy handling and storage Alkali Scientific Flat-Bottom Plates, Non-Treated are available in 48-well.
Catalog No. 50-233-6197 Supplier Alkali Scientific Supplier No. TPN1048NT
Add to Cart
Add to Cart
Use Alkali Scientific Flat-Bottom Plates, Non-Treated for a variety of cell culture applications including suspension cell growth. These sterile plates have a clear, untreated, hydrophobic polystyrene surface, and are ready for use in an aseptic environment. These plates are RNase/DNase free and non-pyrogenic, and the included lid is non-reversible, designed to prevent cross-condensation between wells. Stackable for easy handling and storage Alkali Scientific Flat-Bottom Plates, Non-Treated are available in 48-well.