Learn More
Alkali Scientific CellPro™ 50mL Marker Conical Tubes, Centrifuge Tubes, Conical Bottom with White Screw Caps, Sterile, 25/Resealable Bag, 500/cs
SDP
Catalog No. 502335957
Encompass_Preferred
CellPro™ 50mL Marker Centrifuge Tubes feature a unique white marker screw cap that is easy to write on with a Sharpie or similar lab marker and allows for easy identification of sample on the top. They also have easy-to-read graduations and an oversized white ID patch on the sidewall. The conical tubes are sterile, RNase, DNase and pyrogen free, and come packaged in 20 resealable bags of 25, 500 conical tubes per case. CellPro™ Marker C2entrifuge Tubes are highly resistant to organic solvents and exhibit a stable sample collection and processing environment for nearly all liquids and solids. Tube Temperature range -196°C to +121°C. Cap Temperature range -80°C to +100°C.
Catalog No. 50-233-5957 Supplier Alkali Scientific Supplier No. CW5603
CellPro™ 50mL Marker Centrifuge Tubes feature a unique white marker screw cap that is easy to write on with a Sharpie or similar lab marker and allows for easy identification of sample on the top. They also have easy-to-read graduations and an oversized white ID patch on the sidewall. The conical tubes are sterile, RNase, DNase and pyrogen free, and come packaged in 20 resealable bags of 25, 500 conical tubes per case. CellPro™ Marker C2entrifuge Tubes are highly resistant to organic solvents and exhibit a stable sample collection and processing environment for nearly all liquids and solids. Tube Temperature range -196°C to +121°C. Cap Temperature range -80°C to +100°C.