Learn More
Alkali Scientific CellPro™ 50ML Serological Pipet, Indiv. Wrap, 200/cs
SDP
Catalog No. 502335926
Encompass_Preferred
Serological Pipets • STERILE • Individually Wrapped Serological Pipettes • color coded paper/plastic wrapping for pop-through or peel-apart techniques • Stretched, Non-welded Design • Bi-directional and extra Negative Graduation Marks • Filter Plug and 100% Polystyrene • Gamma Sterilized; Pyrogen/Cytotoxin/RNase & Dnase Free
Catalog No. 50-233-5926 Supplier Alkali Scientific Supplier No. SP250CS
Serological Pipets • STERILE • Individually Wrapped Serological Pipettes • color coded paper/plastic wrapping for pop-through or peel-apart techniques • Stretched, Non-welded Design • Bi-directional and extra Negative Graduation Marks • Filter Plug and 100% Polystyrene • Gamma Sterilized; Pyrogen/Cytotoxin/RNase & Dnase Free