Learn More
Alkali Scientific CellPro™ 500mL Bottle Top Filter, PES Filter Material, 0.45μm, 75mm, Individually Wrapped, 24/cs
SDP
Supplier:  Alkali Scientific VH50045
Encompass_Preferred
CellPro™ Bottle Top Filters are individually wrapped and available in 0.45µm PES membrane. The screw-on funnel includes a lid and a graduated nozzle connection to fit multiple diameter vacuum hoses. They also feature engraved graduation marks to ensure accuracy and are gamma sterilized, certified RNAse, DNAse, sterile and pyrogen free. They offer extremely high flow rates at very low differential pressure. Recommended use is > -60 kPa (kilopascal) and maximum use is > -90 kPa (kilopascal). PES (Polythersulfone) material is proven to be lower protein binding than CA (Calcium Acetate) with far superior durability.
Catalog No. 50-233-6117