Learn More
Alkali Scientific CellPro™ 50mL Flip Top Vial/Digestion Tube, 32×90mm, Self-Standing, Sterile, 500/case
SDP
Catalog No. 503029332
Encompass_Preferred
These new flat bottomed vials are made of natural polypropylene. They feature a one-piece design that includes an integral hinged cap and can be used in digestion equipment / protocols. Tubes are supplied in sterile use-packs of 25 pieces, 500 per case. • Self-standing, flat bottom• Attached caps• Molded graduations• Sterile bags of 25 – 500 per case
Catalog No. 50-302-9332 Supplier Alkali Scientific Supplier No. CT0050
These new flat bottomed vials are made of natural polypropylene. They feature a one-piece design that includes an integral hinged cap and can be used in digestion equipment / protocols. Tubes are supplied in sterile use-packs of 25 pieces, 500 per case. • Self-standing, flat bottom• Attached caps• Molded graduations• Sterile bags of 25 – 500 per case