Learn More
Alkali Scientific CellPro™ 5mL Serological Pipet, Individually Wrapped, (2 X 100) 200/cs
SDP
Catalog No. NC2279910
Encompass_Preferred
CellPro™ Serological Pipets are sterile and individually wrapped. They have color coded paper/plastic wrapping for pop-through or peel-apart techniques. Designed with quality in mind with a stretched, non-welded design and bi-directional and extra negative graduation marks. They contain a filter plug, are 100% polystyrene, and are Gamma sterilized as well as pyrogen, cytotoxin, RNase and Dnase free.
This item is not returnable. View return policy
Catalog No. NC2279910 Supplier Alkali Scientific Supplier No. SP205
CellPro™ Serological Pipets are sterile and individually wrapped. They have color coded paper/plastic wrapping for pop-through or peel-apart techniques. Designed with quality in mind with a stretched, non-welded design and bi-directional and extra negative graduation marks. They contain a filter plug, are 100% polystyrene, and are Gamma sterilized as well as pyrogen, cytotoxin, RNase and Dnase free.