Learn More
Alkali Scientific CellPro™ 7mL Round Bottom Centrifuge Tube with Attached Snap Cap, with Molded Graduations and Frosted Marking Area, Polypropylene, 1000/Pk
SDP
Catalog No. 503026668
Encompass_Preferred
Stop fumbling with caps for your tubes! • Microtube convenience in larger capacities• Attached caps seal at the rim with a “click”• Sterile, easy-open bags of 100 tubes• Molded graduations and frosted marking area   The new round bottom snap cap culture/centrifuge tubes adopt the conveniences of popular 1.5mL microcentrifuge tubes in higher capacities of 5mL, 7mL, and 10mL. Prevent a common source of cross-contamination in the lab by eliminating the threat of mismatched caps. These durable, polypropylene tubes are supplied sterile and are autoclavable. All tubes feature molded graduations and the 5 and 7mL tubes have frosted marking areas. Tubes are packed in easy-to-open sterile bags of 100.
Catalog No. 50-302-6668 Supplier Alkali Scientific Supplier No. SCT907
Stop fumbling with caps for your tubes! • Microtube convenience in larger capacities• Attached caps seal at the rim with a “click”• Sterile, easy-open bags of 100 tubes• Molded graduations and frosted marking area   The new round bottom snap cap culture/centrifuge tubes adopt the conveniences of popular 1.5mL microcentrifuge tubes in higher capacities of 5mL, 7mL, and 10mL. Prevent a common source of cross-contamination in the lab by eliminating the threat of mismatched caps. These durable, polypropylene tubes are supplied sterile and are autoclavable. All tubes feature molded graduations and the 5 and 7mL tubes have frosted marking areas. Tubes are packed in easy-to-open sterile bags of 100.