Learn More
Alkali Scientific CellPro™ 8-Strip Well Tip Combs for Kingfisher™, V-Bottom, PP, 240 bags of 2 pieces, 480/Cs, 8, V-Bottom, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503028775
Encompass_Preferred
CellPro™ 8 Well, V-Bottom, PPTip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc.  • Compatible with KingFisher™ Flex• Low-binding, medical-grade polypropyleneZ• RNase, DNase free
Catalog No. 50-302-8775 Supplier Alkali Scientific Supplier No. TC334008
CellPro™ 8 Well, V-Bottom, PPTip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc.  • Compatible with KingFisher™ Flex• Low-binding, medical-grade polypropyleneZ• RNase, DNase free