Learn More
Alkali Scientific CellPro™ 96 Well Cell Culture Plate, V-Bottom, 1/Sterile inner pack, 100/cs
SDP
Catalog No. 502336177
Encompass_Preferred
CellPro™ 96 Well Cell Culture Plate features a V-bottom design and is non-treated, ensuring compatibility with a wide range of cell types and applications. Each pack is sterile, containing 1 plate, with a case quantity of 100 for bulk purchase convenience. This plate is ideal for cell culture experiments, promoting cell growth and adherence for various research needs. The V-bottom design aids in minimizing the volume of residual liquid left in wells after pipetting, reducing wastage and ensuring precise results. With a well-organized layout of 96 wells, this plate allows for efficient handling of multiple samples simultaneously, saving time and resources in the laboratory. Whether for academic research or industrial testing, this cell culture plate provides a reliable platform for cultivating cells in a controlled environment, facilitating accurate and reproducible experimental outcomes.
Catalog No. 50-233-6177 Supplier Alkali Scientific Supplier No. TPN9094
CellPro™ 96 Well Cell Culture Plate features a V-bottom design and is non-treated, ensuring compatibility with a wide range of cell types and applications. Each pack is sterile, containing 1 plate, with a case quantity of 100 for bulk purchase convenience. This plate is ideal for cell culture experiments, promoting cell growth and adherence for various research needs. The V-bottom design aids in minimizing the volume of residual liquid left in wells after pipetting, reducing wastage and ensuring precise results. With a well-organized layout of 96 wells, this plate allows for efficient handling of multiple samples simultaneously, saving time and resources in the laboratory. Whether for academic research or industrial testing, this cell culture plate provides a reliable platform for cultivating cells in a controlled environment, facilitating accurate and reproducible experimental outcomes.