Learn More
Alkali Scientific CellPro™ 96 Well Cell Culture Plate with Lid, Flat, TC treated, Sterile 10/pk, 100/cs
SDP
Catalog No. 502335797
Encompass_Preferred
CellPro™ cell culture plates have round edges for better handling. Low profile, low-evaporation Lid design assure uniform growth. Choose tissue culture-treated or non treated surfaces. Non treated growth surface is more hydrophobic than tissue culture surface and is similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.
Catalog No. 50-233-5797 Supplier Alkali Scientific Supplier No. TPN2096
CellPro™ cell culture plates have round edges for better handling. Low profile, low-evaporation Lid design assure uniform growth. Choose tissue culture-treated or non treated surfaces. Non treated growth surface is more hydrophobic than tissue culture surface and is similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.