Learn More
Alkali Scientific CellPro™ 96 Well Micro Filtration Plate, 300 µL, for High Throughput Sample Prep, 10/Pk, PVDF, 0.22µm, Hydrophobic
SDP
Catalog No. 5000662533
Encompass_Preferred
The CellPro™ 96 Well Micro Filtration Plate system is engineered for rapid and reliable sample processing in high throughput laboratory workflows. Each plate contains precision bonded membranes with defined pore s such as 0.22 µm or 0.45 µm, enabling efficient clarification, sterile filtration, and particulate removal from a wide variety of biological and chemical samples. With a typical working volume of approximately 300 µL per well, the system supports parallel processing while maintaining uniform flow across wells. The complete four piece configuration includes a filter plate, collection plate, deflector plate, and protective lid, helping prevent cross contamination and ensuring clean filtrate recovery. Designed for compatibility with standard 96 well automation platforms, these plates are ideal for genomic, proteomic, analytical chemistry, and sample preparation workflows prior to techniques such as HPLC, LC MS, or qPCR.
Catalog No. 50-006-62533 Supplier Alkali Scientific Supplier No. FP9622PHOBPVDF
Add to Cart
Add to Cart
The CellPro™ 96 Well Micro Filtration Plate system is engineered for rapid and reliable sample processing in high throughput laboratory workflows. Each plate contains precision bonded membranes with defined pore s such as 0.22 µm or 0.45 µm, enabling efficient clarification, sterile filtration, and particulate removal from a wide variety of biological and chemical samples. With a typical working volume of approximately 300 µL per well, the system supports parallel processing while maintaining uniform flow across wells. The complete four piece configuration includes a filter plate, collection plate, deflector plate, and protective lid, helping prevent cross contamination and ensuring clean filtrate recovery. Designed for compatibility with standard 96 well automation platforms, these plates are ideal for genomic, proteomic, analytical chemistry, and sample preparation workflows prior to techniques such as HPLC, LC MS, or qPCR.