Learn More
Alkali Scientific CellPro™ 96 Well Micro Filtration Plate, 600 µL, PTEF, Hydrophilic, for High Throughput Sample Prep, 10/Pk, 0.45µm
SDP
Catalog No. 5000662320
Encompass_Preferred
The CellPro™ 96 Well Micro Filtration Plate is designed for high volume, high throughput sample preparation in molecular biology, analytical chemistry, and drug discovery workflows. Each well supports up to 600 µL capacity, allowing larger sample loads and improved recovery compared with standard filtration plates. Precision engineered membranes with defined pore s provide effective clarification, sterile filtration, and removal of particulates from a wide range of biological and chemical samples. The complete four piece assembly includes a filter plate, collection plate, deflector plate, and protective lid to ensure clean separation and minimize cross contamination. Optimized for both vacuum and centrifuge filtration, the plate integrates seamlessly with standard 96 well automation systems, making it ideal for screening laboratories, high throughput purification, lysate cleanup, and advanced sample preparation workflows.
Catalog No. 50-006-62320 Supplier Alkali Scientific Supplier No. FP09645
Add to Cart
Add to Cart
The CellPro™ 96 Well Micro Filtration Plate is designed for high volume, high throughput sample preparation in molecular biology, analytical chemistry, and drug discovery workflows. Each well supports up to 600 µL capacity, allowing larger sample loads and improved recovery compared with standard filtration plates. Precision engineered membranes with defined pore s provide effective clarification, sterile filtration, and removal of particulates from a wide range of biological and chemical samples. The complete four piece assembly includes a filter plate, collection plate, deflector plate, and protective lid to ensure clean separation and minimize cross contamination. Optimized for both vacuum and centrifuge filtration, the plate integrates seamlessly with standard 96 well automation systems, making it ideal for screening laboratories, high throughput purification, lysate cleanup, and advanced sample preparation workflows.