Learn More
Alkali Scientific CellPro™ 96 Well Round Bottom Cell Culture Plates with Lid, TC Treated, 100/pk
SDP
Catalog No. 502336141
Encompass_Preferred
CellPro™ cell culture plates have round edges for better handling. Low profile, low-evaporation Lid design assure uniform growth. Choose tissue culture-treated or non treated surfaces. Non treated growth surface is more hydrophobic than tissue culture surface and is similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.
Catalog No. 50-233-6141 Supplier Alkali Scientific Supplier No. TPN9095
Add to Cart
Add to Cart
CellPro™ cell culture plates have round edges for better handling. Low profile, low-evaporation Lid design assure uniform growth. Choose tissue culture-treated or non treated surfaces. Non treated growth surface is more hydrophobic than tissue culture surface and is similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.