Learn More
Alkali Scientific CellPro™ 96-Well, Silicone Sealing Mats, 50/pk, 96 Round Well, Black Alphanumeric Printing, Silicone, "+" Opening, Ø7.3mm, Printed 50 / PK
SDP
Catalog No. 503027764
Encompass_Preferred
CellPro™ 96 Well, Silicone Sealing Mats, 50/pk mat seals 96 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.
Catalog No. 50-302-7764 Supplier Alkali Scientific Supplier No. SSM332013
Add to Cart
Add to Cart
CellPro™ 96 Well, Silicone Sealing Mats, 50/pk mat seals 96 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.