Learn More
Alkali Scientific CellPro™ 96-Well Cell Culture Plate, Flat, Non-Treated, Sterile, 1/pk, 100/cs
SDP
Catalog No. 502335863
Encompass_Preferred
CellPro™ cell culture plates have round edges for better handling. Low profile and low-evaporation lid design assures uniform growth. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.
Catalog No. 50-233-5863 Supplier Alkali Scientific Supplier No. TPN32
CellPro™ cell culture plates have round edges for better handling. Low profile and low-evaporation lid design assures uniform growth. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.