Learn More
Alkali Scientific CellPro™ ACK Lysing, 1L
SDP
Catalog No. 502389290
Encompass_Preferred
Alkali Scientific™ proudly introduces CellPro™ ACK Lysing Buffer, a sterile solution designed for efficient red blood cell lysis in biological samples. This specialized buffer ensures the gentle and thorough removal of erythrocytes, facilitating downstream analysis in flow cytometry, cell sorting, and other hematological applications. With its sterile formulation, CellPro™ ACK Lysing Buffer maintains sample integrity and minimizes the risk of contamination, ensuring reliable and reproducible results. Streamline your cell analysis workflows with the high-quality performance and convenience of CellPro™ ACK Lysing Buffer from Alkali Scientific™.
Catalog No. 50-238-9290 Supplier Alkali Scientific Supplier No. DB0309
Alkali Scientific™ proudly introduces CellPro™ ACK Lysing Buffer, a sterile solution designed for efficient red blood cell lysis in biological samples. This specialized buffer ensures the gentle and thorough removal of erythrocytes, facilitating downstream analysis in flow cytometry, cell sorting, and other hematological applications. With its sterile formulation, CellPro™ ACK Lysing Buffer maintains sample integrity and minimizes the risk of contamination, ensuring reliable and reproducible results. Streamline your cell analysis workflows with the high-quality performance and convenience of CellPro™ ACK Lysing Buffer from Alkali Scientific™.