Learn More
Alkali Scientific CellPro™ Agar Biological Grade 4% 500mL
SDP
Catalog No. 502388832
Encompass_Preferred
CellPro™ Agar Biological Grade is a high-quality agar medium designed for microbiological and molecular biology applications in laboratory settings. This agar powder is derived from select seaweed sources and undergoes stringent quality control processes to ensure purity and consistency. Biological Grade Agar is commonly used to prepare solid culture media for the cultivation and isolation of microorganisms, including bacteria, yeast, and fungi. It provides a solid substrate for microbial growth and is suitable for a wide range of microbiological techniques, such as colony counting, streaking, and microbial identification.
Catalog No. 50-238-8832 Supplier Alkali Scientific Supplier No. DC42240
Add to Cart
Add to Cart
CellPro™ Agar Biological Grade is a high-quality agar medium designed for microbiological and molecular biology applications in laboratory settings. This agar powder is derived from select seaweed sources and undergoes stringent quality control processes to ensure purity and consistency. Biological Grade Agar is commonly used to prepare solid culture media for the cultivation and isolation of microorganisms, including bacteria, yeast, and fungi. It provides a solid substrate for microbial growth and is suitable for a wide range of microbiological techniques, such as colony counting, streaking, and microbial identification.