Learn More
Alkali Scientific CellPro™ Agarose Gel Loading Dye, 6X Glycerol Based, 25mL
SDP
Catalog No. 502389192
Encompass_Preferred
CellPro™ Gel Loading Dye is a specialized solution used in gel electrophoresis procedures for visualizing DNA or RNA samples during analysis. This loading dye contains tracking dyes to aid in monitoring the progress of electrophoresis and ensuring accurate loading of samples into the gel wells. Its formulation helps to enhance sample visibility and migration, facilitating precise analysis and interpretation of results.
Catalog No. 50-238-9192 Supplier Alkali Scientific Supplier No. AG6001
CellPro™ Gel Loading Dye is a specialized solution used in gel electrophoresis procedures for visualizing DNA or RNA samples during analysis. This loading dye contains tracking dyes to aid in monitoring the progress of electrophoresis and ensuring accurate loading of samples into the gel wells. Its formulation helps to enhance sample visibility and migration, facilitating precise analysis and interpretation of results.