Learn More
Alkali Scientific CellPro Amber Centrifuge Tubes
SDP
Supplier:  Alkali Scientific CW5602AMB
Encompass_Preferred
The CellProAmber Centrifuge Tubes are engineered for superior sample protection and performance Constructed from durable polypropylene (PP) these conical tubes feature an amber design to safeguard light-sensitive materials by significantly reducing light penetration Each tube is paired with a high-density polyethylene (HDPE) cap ensuring a secure leak-proof seal for reliable sample containment Amber Design for Light Protection Shields light-sensitive samples from degradation by minimizing light transmission HDPE Caps High-density polyethylene caps provide a tight secure fit to prevent leaks and maintain sample integrity during storage and centrifugation High Centrifugal Strength Robust design supports high-speed centrifugation with maximum RCF ratings of 13000 x g for 15 mL tubes and 14000 x g for 50 mL tubes Chemical Resistance Made from chemically resistant materials suitable for a variety of applications Bulk Packaging Conveniently packed with 500 tubes per case to meet the n
Catalog No. 50-302-7198