Learn More
Alkali Scientific CellPro Amber Centrifuge Tubes
SDP
Supplier:  Alkali Scientific CW5601AMB
Encompass
The CellProAmber Centrifuge Tubes are engineered for superior sample protection and performance Constructed from durable polypropylene (PP) these conical tubes feature an amber design to safeguard light-sensitive materials by significantly reducing light penetration Each tube is paired with a high-density polyethylene (HDPE) cap ensuring a secure leak-proof seal for reliable sample containment Amber Design for Light Protection Shields light-sensitive samples from degradation by minimizing light transmission HDPE Caps High-density polyethylene caps provide a tight secure fit to prevent leaks and maintain sample integrity during storage and centrifugation High Centrifugal Strength Robust design supports high-speed centrifugation with maximum RCF ratings of 13000 x g for 15 mL tubes and 14000 x g for 50 mL tubes Chemical Resistance Made from chemically resistant materials suitable for a variety of applications Bulk Packaging Conveniently packed with 500 tubes per case to meet the n
Catalog No. 50-302-7283