Learn More
Alkali Scientific CellPro™ Ammonium Sulfate, 1M, 100mL
SDP
Catalog No. 502389411
Encompass_Preferred
Alkali Scientific™ CellPro™ Ammonium Sulfate, 1M, offers a meticulously prepared solution for various molecular biology and biochemical applications. This sterile and filtered solution ensures exceptional purity and reliability, meeting the stringent demands of laboratory protocols. Ammonium sulfate is a commonly used reagent in laboratory settings, employed for processes such as protein precipitation, enzyme crystallization, and nucleic acid isolation. With its carefully controlled formulation and rigorous quality control measures, CellPro™ Ammonium Sulfate guarantees consistent and reproducible results, empowering researchers to achieve their scientific goals with confidence. Trust in the purity and reliability of CellPro™ Ammonium Sulfate for your laboratory's molecular biology and biochemical requirements.
Catalog No. 50-238-9411 Supplier Alkali Scientific Supplier No. DB0237
Alkali Scientific™ CellPro™ Ammonium Sulfate, 1M, offers a meticulously prepared solution for various molecular biology and biochemical applications. This sterile and filtered solution ensures exceptional purity and reliability, meeting the stringent demands of laboratory protocols. Ammonium sulfate is a commonly used reagent in laboratory settings, employed for processes such as protein precipitation, enzyme crystallization, and nucleic acid isolation. With its carefully controlled formulation and rigorous quality control measures, CellPro™ Ammonium Sulfate guarantees consistent and reproducible results, empowering researchers to achieve their scientific goals with confidence. Trust in the purity and reliability of CellPro™ Ammonium Sulfate for your laboratory's molecular biology and biochemical requirements.