Learn More
Alkali Scientific CellPro™ Amphotericin B (Fungiezone) SOLN 250 ug/mL, 50mL, 50ml
SDP
Supplier:  Alkali Scientific DCC008
Encompass_Preferred
An antifungal, anti-yeast and anti-mold reagent. An antibiotic agent that is active against fungi Interferes with the permeability of the cell membrane of sensitive fungi by binding sterols Common working concentration is 2.5 μg/mL, and it is cytotoxic at 30 μg/mL Used as an antifungal, anti-yeast and anti-mold agent
Catalog No. 50-302-7284