Learn More
Alkali Scientific CellPro™ Amphotericin B (Fungiezone) SOLN 250 ug/mL, 50mL, 50ml, 10/Case
SDP
Catalog No. 503026556
Encompass_Preferred
An antifungal, anti-yeast and anti-mold reagent. An antibiotic agent that is active against fungi Interferes with the permeability of the cell membrane of sensitive fungi by binding sterols Common working concentration is 2.5 μg/mL, and it is cytotoxic at 30 μg/mL Used as an antifungal, anti-yeast and anti-mold agent
Catalog No. 50-302-6556 Supplier Alkali Scientific Supplier No. DCC008CS
An antifungal, anti-yeast and anti-mold reagent. An antibiotic agent that is active against fungi Interferes with the permeability of the cell membrane of sensitive fungi by binding sterols Common working concentration is 2.5 μg/mL, and it is cytotoxic at 30 μg/mL Used as an antifungal, anti-yeast and anti-mold agent