Learn More
Alkali Scientific CellPro™ Aspirating Pipet, 5mL, 200/Pk, (1 Self Standing Inner Box)
SDP
Catalog No. 503028967
Encompass_Preferred
These non-plugged, nongraduated, single-use pipet can be used for a wide variety of vacuum-aspirating procedures, and are a safer alternative to glass pasteur pipets. Pipets are supplied in a thermoformed paper/plastic packaging wrap to protect against contamination. Nonpyrogenic. • eBeam STERILE• 5mL only with other sizes available upon request• Supplied without graduations or plugs• Individually Wrapped• Each Pipette Packaging: single wrapped STERILE, paper/plastic wrappers allow easy Punch thru -or- Peel to open
Catalog No. 50-302-8967 Supplier Alkali Scientific Supplier No. SP205A
These non-plugged, nongraduated, single-use pipet can be used for a wide variety of vacuum-aspirating procedures, and are a safer alternative to glass pasteur pipets. Pipets are supplied in a thermoformed paper/plastic packaging wrap to protect against contamination. Nonpyrogenic. • eBeam STERILE• 5mL only with other sizes available upon request• Supplied without graduations or plugs• Individually Wrapped• Each Pipette Packaging: single wrapped STERILE, paper/plastic wrappers allow easy Punch thru -or- Peel to open