Learn More
Alkali Scientific CellPro™ Automation Reservoir Cover, Polypropylene, 200/Case, STERILE
SDP
Catalog No. 503028611
Encompass_Preferred
OptiWell" Reservoirs are compatible with a wide range of automated liquid handling equipment. Offered with 1 to 384 wells, they are useful in multiple applications, including mixing, fraction collection, plate replication, compound storage, and scale-up / scale-down methods. OptiWell" Reservoirs conform with SLAS standard dimensions. They are made of chemically inert, low binding polypropylene to minimize excess residue/waste. Certified RNase, DNase and pyrogen free. Features: ANSI / SLAS 1-2004 Compliant  Standard footprint dimensions Nonpyrogenic, RNase, DNase free Low-binding polypropylene
Catalog No. 50-302-8611 Supplier Alkali Scientific Supplier No. RS3390CVS
OptiWell" Reservoirs are compatible with a wide range of automated liquid handling equipment. Offered with 1 to 384 wells, they are useful in multiple applications, including mixing, fraction collection, plate replication, compound storage, and scale-up / scale-down methods. OptiWell" Reservoirs conform with SLAS standard dimensions. They are made of chemically inert, low binding polypropylene to minimize excess residue/waste. Certified RNase, DNase and pyrogen free. Features: ANSI / SLAS 1-2004 Compliant  Standard footprint dimensions Nonpyrogenic, RNase, DNase free Low-binding polypropylene