Learn More
Alkali Scientific CellPro™ Automation Reservoirs, 195ml, 4 Channel, Polypropylene, Universally Robotic Liquid Handler Compatible, 50/Case, STERILE
SDP
Catalog No. 503028454
Encompass_Preferred
Robotic liquid handler compatible ANSI / SLAS 1-2004 Compliant  Standard footprint dimensions Nonpyrogenic, RNase, DNase free Low-binding polypropylene ISO Class 8 Cleanroom Certified These automaton reagent solution reservoirs are designed to be compatible with all automated liquid handling platforms. Dimpled and pyramid-shaped well bottoms assist in maximum sample recovery. Offered with 1 to 384 wells, they are useful in multiple applications, including mixing, fraction collection, plate replication, compound storage, and scale-up / scale-down methods.
Catalog No. 50-302-8454 Supplier Alkali Scientific Supplier No. RS339004S
Robotic liquid handler compatible ANSI / SLAS 1-2004 Compliant  Standard footprint dimensions Nonpyrogenic, RNase, DNase free Low-binding polypropylene ISO Class 8 Cleanroom Certified These automaton reagent solution reservoirs are designed to be compatible with all automated liquid handling platforms. Dimpled and pyramid-shaped well bottoms assist in maximum sample recovery. Offered with 1 to 384 wells, they are useful in multiple applications, including mixing, fraction collection, plate replication, compound storage, and scale-up / scale-down methods.