Learn More
Alkali Scientific CellPro™ BenchBin™ Biohazard Waste Container, Benchtop Biohazard Bin, 3L
SDP
Supplier:  Alkali Scientific A3404B
Encompass_Preferred
BenchBin™ is a personal Biohazard waste container that any researcher can benefit from. Use with 8.5 x 11in. autoclave bags to contain biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous materials. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat. Safely dispose of tips, tubes, gloves, etc. Secures and covers hazardous waste Easy push open/close spring lid Compact footprint – 7 x 5 x 7in.
Catalog No. 50-302-6604