Learn More
Alkali Scientific CellPro™ BenchBin™ Biohazard Waste Container, Starter Kit Includes 1 bin and 400 autoclave bags (8.5 x 11in.)
SDP
Catalog No. 503026416
Encompass_Preferred
BenchBin™ is a personal Biohazard waste container that any researcher can benefit from. Use with 8.5 x 11in. autoclave bags to contain biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous materials. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat. Safely dispose of tips, tubes, gloves, etc. Secures and covers hazardous waste Easy push open/close spring lid Compact footprint – 7 x 5 x 7in.
Catalog No. 50-302-6416 Supplier Alkali Scientific Supplier No. A3001SK
Add to Cart
Add to Cart
BenchBin™ is a personal Biohazard waste container that any researcher can benefit from. Use with 8.5 x 11in. autoclave bags to contain biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous materials. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, the BenchBin features a push to open/close lid that keeps materials covered and your bench neat. Safely dispose of tips, tubes, gloves, etc. Secures and covers hazardous waste Easy push open/close spring lid Compact footprint – 7 x 5 x 7in.