Learn More
Alkali Scientific CellPro™ Blender bag, 400ml x 70µm with full nylon filter
SDP
Catalog No. 503029039
Encompass_Preferred
Blender bag, LPDE, 400ml x 70µmwith full nylon filter, 30 x 19cm, pack of 500 (10 packs of 50)
Catalog No. 50-302-9039 Supplier Alkali Scientific Supplier No. RS20523
Blender bag, LPDE, 400ml x 70µmwith full nylon filter, 30 x 19cm, pack of 500 (10 packs of 50)