Learn More
Alkali Scientific CellPro™ Blender bag, with lateral (side) nylon filter
SDP
Catalog No. 503027713
Encompass_Preferred
Blender bag, LPDE, 400ml x 70µm with lateral (side) nylon filter, 30 x 19cm, pack of 500 (10 packs of 50)
Catalog No. 50-302-7713 Supplier Alkali Scientific Supplier No. RS20522
Blender bag, LPDE, 400ml x 70µm with lateral (side) nylon filter, 30 x 19cm, pack of 500 (10 packs of 50)